Learn More
Grainger Cooling Bandana PVA And Cotton One Size
Catalog No. 502767725
Encompass_Preferred
The Chill-Its by Ergodyne Evaporative Cooling Bandana is a reusable cooling bandana that provides up to 30 minutes of cooling relief. It is made of hyper-evaporative material that activates when wet to provide a cooling effect. To use, soak in water for 1-2 minutes then wring out excess. It will stay cool even during strenuous activity. The bandana is 12" x 24" in size and machine washable. It is lightweight, breathable, and comfortable to wear around neck or head. The cooling bandana is ideal for working in hot conditions to prevent overheating.
Catalog No. 50-276-7725 Supplier Grainger Supplier No. 16V829
The Chill-Its by Ergodyne Evaporative Cooling Bandana is a reusable cooling bandana that provides up to 30 minutes of cooling relief. It is made of hyper-evaporative material that activates when wet to provide a cooling effect. To use, soak in water for 1-2 minutes then wring out excess. It will stay cool even during strenuous activity. The bandana is 12" x 24" in size and machine washable. It is lightweight, breathable, and comfortable to wear around neck or head. The cooling bandana is ideal for working in hot conditions to prevent overheating.