Learn More
Electron Microscopy Sciences CoolSafe™ Cooling Chamber for 1.5 ml tubes
SDP
Catalog No. 5033420
Encompass_Preferred
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.
Catalog No. 50-334-20 Supplier Electron Microscopy Sciences Supplier No. 6195130
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.