Learn More
Electron Microscopy Sciences Cooling Chamber for 50 ml tubes, 4 holes
SDP
Catalog No. 5033424
Encompass_Preferred
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe.
Catalog No. 50-334-24 Supplier Electron Microscopy Sciences Supplier No. 6195541
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe.