Learn More
Electron Microscopy Sciences Cooling Chamber for 15 ml tubes, 6 holes
SDP
Catalog No. 5033427
Encompass_Preferred
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe.
Catalog No. 50-334-27 Supplier Electron Microscopy Sciences Supplier No. 6195520
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe.