Learn More
Electron Microscopy Sciences CoolSafe™ Cooling Chamber for multi-tube format
SDP
Catalog No. 5033423
Encompass_Preferred
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.
Catalog No. 50-334-23 Supplier Electron Microscopy Sciences Supplier No. 6195170
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.