Learn More
Bellco Glass, Inc. Cooling Coil Assembly , 15L
SDP
Supplier:  Bellco Glass, Inc. 790915007
Encompass_Preferred
For 18mm portMaterial 316l sst with ep finish of ra 32 or better - outside onlyWith hose barbs on each endApprox 11 coilsTubing specs 1/4 od x 5/32" idDimensions: 17" overall length with 4.468" center one tube - center of other tubeInsertion length 15 1/2"Supplied with A555-23 - washer/A555-3-nut & A523-234 - o-ring. MADE IN THE USA
Catalog No. 50-194-6476