Learn More
Julabo USA Inc Cooling coil for PURA 10, PURA 14
SDP
Catalog No. 501904636
Encompass_Preferred
Cooling coil for use with PURA 10 and PURA 14 water baths for operation near ambient temperature. Connect to cooling water for operation down to +18 ¿C.
Catalog No. 50-190-4636 Supplier Julabo USA Inc Supplier No. 9970534
Cooling coil for use with PURA 10 and PURA 14 water baths for operation near ambient temperature. Connect to cooling water for operation down to +18 ¿C.