Learn More
Julabo USA Inc Cooling coil for PURA 22, PURA 30
SDP
Catalog No. 501904637
Encompass_Preferred
Cooling coil for use with PURA 22 and PURA 30 water baths for operation near ambient temperature. Connect to cooling water for operation down to +18 ¿C.
Catalog No. 50-190-4637 Supplier Julabo USA Inc Supplier No. 9970536
Cooling coil for use with PURA 22 and PURA 30 water baths for operation near ambient temperature. Connect to cooling water for operation down to +18 ¿C.