Learn More
Bellco Glass, Inc. Cooling Finger Assembly , 3L
SDP
Catalog No. 501946425
Encompass_Preferred
Stainless steel cooling finger with internally placed water in and water out tubes that help to maintain internal bioreactor temperature.--Fits into 18mm port on 3Lmetric headplate--316L Stainless Steel--Electropolish Finish. MADE IN THE USA
Catalog No. 50-194-6425 Supplier Bellco Glass, Inc. Supplier No. 790900011
Stainless steel cooling finger with internally placed water in and water out tubes that help to maintain internal bioreactor temperature.--Fits into 18mm port on 3Lmetric headplate--316L Stainless Steel--Electropolish Finish. MADE IN THE USA