Learn More
Bellco Glass, Inc. Cooling Finger Assembly , 7L
SDP
Catalog No. 501946445
Encompass_Preferred
Stainless steal cooling finger with internally placed water in and water out tubes that help to maintain internal bioreactor temperature. Fits into 18mm port on 3L metric headplate. 316L SST. MADE IN THE USA
Catalog No. 50-194-6445 Supplier Bellco Glass, Inc. Supplier No. 790900036
Stainless steal cooling finger with internally placed water in and water out tubes that help to maintain internal bioreactor temperature. Fits into 18mm port on 3L metric headplate. 316L SST. MADE IN THE USA