Learn More
Cole-Parmer Techne BLoCKICE Cooling/Heating Block, 100-230 VAC, 50/60 Hz
Catalog No. 501960487 Shop All Cole-Parmer Products
Encompass
This benchtop chilling and heating block is ideal for incubating sample tubes at temperatures ranging from 32 to 104°F (0 to 40°C). The Isotemptemperature control ensures all contents are kept at an even temperature, whether cold or warm. Aluminum block inserts provide a temperature stability of±0.2°C for precise control of reaction temperature. Unit holds two single-width inserts or one double-width inserts. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached. A dark plastic lid protects light-sensitive reagents while maintaining the cooled environment.
Catalog No. 50-196-0487 Supplier Cole-Parmer Supplier No. 3662017
This benchtop chilling and heating block is ideal for incubating sample tubes at temperatures ranging from 32 to 104°F (0 to 40°C). The Isotemptemperature control ensures all contents are kept at an even temperature, whether cold or warm. Aluminum block inserts provide a temperature stability of±0.2°C for precise control of reaction temperature. Unit holds two single-width inserts or one double-width inserts. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached. A dark plastic lid protects light-sensitive reagents while maintaining the cooled environment.