Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ 96 Storage Tube Modules
BIOPHARMA

Catalog No. 07210057 Shop All Corning Products
Change view
Click to view available options
Holds:
96 x 0.5 mL 2D Bar Code Tubes
96 x 1.4 mL 2D Bar Code Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-210-057 96 x 1.4 mL 2D Bar Code Tubes
07-210-056 96 x 0.5 mL 2D Bar Code Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-210-057 Supplier Corning™ Supplier No. 432057
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • 2D Barcode storage tube modules
  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.
TRUSTED_SUSTAINABILITY

Specifications

Holds 96 x 1.4 mL 2D Bar Code Tubes
Product Type Storage Tube Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.