Learn More
Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
BIOPHARMA
Catalog No. 07210052 Shop All Corning Products
Change view
Click to view available options
Holds:
15 Cryogenic Vials or FACS Tubes
24 Cryogenic Vials or FACS Tubes
30 Cryogenic Vials or FACS Tubes
45 Cryogenic Vials or FACS Tubes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-210-052 30 Cryogenic Vials or FACS Tubes
07-210-049 15 Cryogenic Vials or FACS Tubes
07-210-050 24 Cryogenic Vials or FACS Tubes
07-210-051 45 Cryogenic Vials or FACS Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 07-210-052 Supplier Corning™ Supplier No. 432052
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • Ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY
Specifications
Holds 30 Cryogenic Vials or FACS Tubes
Design CFT30, with gripping wells for one-hand 1 vial opening/closing
Product Type Cryogenic Vial, FACS Tube Module