Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ LX50, for use with Axygen 50mL Reagent Reservoir Module
BIOPHARMA

Catalog No. 07202451 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Axygen 50 mL Reagent Reservoir Module
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-202-451 Axygen 50 mL Reagent Reservoir Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-202-451 Supplier Corning™ Supplier No. 432139
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corning™'s CoolRack™ thermo-conductive reservoir modules provide uniform temperature to all wells, regardless of position.

Corning™ CoolRack™, thermo-conductive reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolRack module will rapidly adapt to that temperature - from -196°C to >+100°C.

  • CoolRack modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • For use with the 50 mL Axygen™ disposable reservoirs.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Axygen 50 mL Reagent Reservoir Module
Quantity 1
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.