Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ M Microcentrifuge Tube Modules
BIOPHARMA

Catalog No. 07210043 Shop All Corning Products
Change view
Click to view available options
Holds:
15 x 1.5 or 2 mL MCT Tubes
24 x 1.5 or 2 mL MCT Tubes, SBS-compatible
30 x 1.5 or 2 mL MCT Tubes
6 x 1.5 or 2 mL MCT Tubes
90 x 1.5 or 2 mL MCT Tubes
96 x 1.5 or 2 mL MCT Tubes with A to H and 1 to 12
Color:
Gray
Green
Orange
Design:
M6
M15
M24
M30
M90
M96
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Color Design
07-210-043 30 x 1.5 or 2 mL MCT Tubes Orange M30
07-210-034 6 x 1.5 or 2 mL MCT Tubes Gray M6
07-210-035 6 x 1.5 or 2 mL MCT Tubes Green M6
07-210-036 6 x 1.5 or 2 mL MCT Tubes Orange M6
07-210-037 15 x 1.5 or 2 mL MCT Tubes Gray M15
07-210-038 15 x 1.5 or 2 mL MCT Tubes Green M15
07-210-039 15 x 1.5 or 2 mL MCT Tubes Orange M15
07-210-040 24 x 1.5 or 2 mL MCT Tubes, SBS-compatible Gray M24
07-210-041 30 x 1.5 or 2 mL MCT Tubes Gray M30
07-210-042 30 x 1.5 or 2 mL MCT Tubes Green M30
07-210-044 90 x 1.5 or 2 mL MCT Tubes Gray M90
07-210-045 96 x 1.5 or 2 mL MCT Tubes with A to H and 1 to 12 Gray M96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 07-210-043 Supplier Corning™ Supplier No. 432043
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo-conductive microfuge tube modules that eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources.

Corning CoolRack thermo-conductive microcentrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
For Use With (Application) Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen
For Use With (Equipment) Microcentrifuge Tubes
Holds 30 x 1.5 or 2 mL MCT Tubes
Array 5 x 6
Design M30
Product Type Microcentrifuge Tube Modules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.