Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ M-PF Conical Tube Modules
BIOPHARMA

Catalog No. 07210048 Shop All Corning Products
Change view
Click to view available options
Holds:
15 x 1.5 mL conical tubes
30 x 1.5 mL Conical Tubes
30 x 500μL Conical Tubes
Design:
M15-PF
M30-PF
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Design
07-210-048 30 x 1.5 mL Conical Tubes M30-PF
07-210-046 30 x 500μL Conical Tubes M30-PF
07-210-047 15 x 1.5 mL conical tubes M15-PF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-210-048 Supplier Corning™ Supplier No. 432048
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive microfuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Holds 30 x 1.5 mL Conical Tubes
Design M30-PF
Product Type Conical Tube Thermoconductive Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.