Learn More
Corning™ CoolRack™ Plate and Reservoir Modules
BIOPHARMAGSA_VA
Catalog No. 07210070 Shop All Corning Products
Change view
Click to view available options
Holds:
1 Flat Bottom Plate, 1 SBS-compatible
1 U-bottom Microplate, SBS compatible
Reagent Reservoir (55 mL)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-210-070 1 Flat Bottom Plate, 1 SBS-compatible
07-210-071 1 U-bottom Microplate, SBS compatible
07-210-072 Reagent Reservoir (55 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-210-070 Supplier Corning™ Supplier No. 432070
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • SBS-compatible
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Holds 1 Flat Bottom Plate, 1 SBS-compatible
Design CoolSink XT 96F
Type CoolSink Plate and Reservoir Module
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov