Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ Plate and Reservoir Modules
BIOPHARMA

Catalog No. 07210072 Shop All Corning Products
Change view
Click to view available options
Holds:
1 Flat Bottom Plate, 1 SBS-compatible
1 U-bottom Microplate, SBS compatible
Reagent Reservoir (55 mL)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-210-072 Reagent Reservoir (55 mL)
07-210-070 1 Flat Bottom Plate, 1 SBS-compatible
07-210-071 1 U-bottom Microplate, SBS compatible
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-210-072 Supplier Corning™ Supplier No. 432072
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.

  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • SBS-compatible
TRUSTED_SUSTAINABILITY

Specifications

Holds Reagent Reservoir (55 mL)
Design CoolSink LX55
Product Type CoolSink Plate and Reservoir Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.