Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolRack™ Tall Modules
BIOPHARMAGSA_VA

Catalog No. 07210062 Shop All Corning Products
Change view
Click to view available options
Holds:
1 250 mL Centrifuge Tube
1 250 mL Easy Grip Storage Bottle
12 x 15 mL Centrifuge Tubes
12 x 5 mL Centrifuge Tubes, SBS-compatible
4 x 50 mL Centrifuge Tubes
9 x 15 mL Centrifuge Tubes
Design:
CoolRack 15 mL
CoolRack 250 mL
CoolRack 250 mL PF
CoolRack 50 mL
CoolRack L
CoolRack XT 5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Design
07-210-062 4 x 50 mL Centrifuge Tubes CoolRack 50 mL
07-210-060 12 x 5 mL Centrifuge Tubes, SBS-compatible CoolRack XT 5 mL
07-210-061 9 x 15 mL Centrifuge Tubes CoolRack 15 mL
07-210-063 1 250 mL Centrifuge Tube CoolRack 250 mL PF
07-210-064 1 250 mL Easy Grip Storage Bottle CoolRack 250 mL
07-210-068 12 x 15 mL Centrifuge Tubes CoolRack L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 07-210-062 Supplier Corning™ Supplier No. 432062
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Holds 4 x 50 mL Centrifuge Tubes
Quantity 1/cs.
Design CoolRack 50 mL
Product Type Tall Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.