Learn More
Corning™ CoolRack™ XT PCR Tube Modules
BIOPHARMA
Catalog No. 07210055 Shop All Corning Products
Change view
Click to view available options
Holds:
1.5 to 2 mL Tubes
384-well PCR Plate
96-well PCR Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07-210-055 384-well PCR Plate
07-210-053 96-well PCR Plate
07-210-054 1.5 to 2 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-210-055 Supplier Corning™ Supplier No. 432055
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • CoolRack modules ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible.
TRUSTED_SUSTAINABILITY
Specifications
Description Capacity: 384-well PCR microplate, SBS-compatible
Holds 384-well PCR Plate
Design PCR384
Product Type PCR Plate Module