Learn More
USA Scientific Inc CoolSafe chamber for 0.2 mL tubes
Catalog No. NC9929933
Encompass
CoolSafe chamber for 0.2 mL tubes or plates, aluminum. Holds 96 0.2 mL tubes, four 1.5 mL tubes, and six 0.5 mL tubes.
Catalog No. NC9929933 Supplier USA Scientific Inc Supplier No. 91240100
CoolSafe chamber for 0.2 mL tubes or plates, aluminum. Holds 96 0.2 mL tubes, four 1.5 mL tubes, and six 0.5 mL tubes.