Learn More
USA Scientific Inc CoolSafe Chamber for 1.5 mL tubes
Catalog No. NC9929931
Encompass
CoolSafe chamber for 1.5 mL tubes, aluminum. Holds 48 1.5 mL tubes.
Catalog No. NC9929931 Supplier USA Scientific Inc Supplier No. 91243000
CoolSafe chamber for 1.5 mL tubes, aluminum. Holds 48 1.5 mL tubes.