Learn More
Electron Microscopy Sciences CoolSafe™ CoolSafe Box
SDP
Catalog No. 5033415
Encompass_Preferred
The CoolSafe Box is made of triple density expanded polystyrene and is molded under extreme pressure to create a durable container that guarantees temperature stability. The CoolBag is an insulated cooler for added coolness of critical samples.  Use the shoulder strap for easy transporting from lab to lab. 
Catalog No. 50-334-15 Supplier Electron Microscopy Sciences Supplier No. 6195301
The CoolSafe Box is made of triple density expanded polystyrene and is molded under extreme pressure to create a durable container that guarantees temperature stability. The CoolBag is an insulated cooler for added coolness of critical samples.  Use the shoulder strap for easy transporting from lab to lab. 