Learn More
Electron Microscopy Sciences CoolSafe™ Kit for flat bottom microplates
SDP
Catalog No. 5033503
Encompass_Preferred
The CoolSafe Deluxe provides additional sample protection, smaller bench-top footprint, imrpoved portability, and clear lid for sample visibility. Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber and 2 Cool Bricks (1 degree C & –10 degree C). These Chambers also include 4 holes for 1.5 ml tubes and 6 holes for 0.5 ml tubes.
Catalog No. 50-335-03 Supplier Electron Microscopy Sciences Supplier No. 6195440
The CoolSafe Deluxe provides additional sample protection, smaller bench-top footprint, imrpoved portability, and clear lid for sample visibility. Each complete CoolSafe Deluxe system includes 1 CoolSafe Deluxe box, 1 Cool Chamber and 2 Cool Bricks (1 degree C & –10 degree C). These Chambers also include 4 holes for 1.5 ml tubes and 6 holes for 0.5 ml tubes.