Learn More
Electron Microscopy Sciences CoolSafe™ Bench Top Cooling System for 5 ml tubes, 32 Capacity
SDP
Catalog No. 5033409
Encompass_Preferred
The coolsafe is a versatile and effective benchtop cooling and storage system designed to hold tubes (strips or trays) and microplates. Specially formulated cool bricks maintain a constant temperature (1 degree C or -10 degree C) for up to 10 hours. Each aluminum chamber surrounds tubes to guarantee sustained cold temperatures. Box is made of triple density expanded polystyrene and molded under extreme pressure creating a durable container that guarantees temperature stability. Easily interchange cool chambers and cool bricks for different sized tubes.
Each complete CoolSafe System includes 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1 degree C & -10 degree C).
Capacity: 32
Catalog No. 50-334-09 Supplier Electron Microscopy Sciences Supplier No. 6195060
The coolsafe is a versatile and effective benchtop cooling and storage system designed to hold tubes (strips or trays) and microplates. Specially formulated cool bricks maintain a constant temperature (1 degree C or -10 degree C) for up to 10 hours. Each aluminum chamber surrounds tubes to guarantee sustained cold temperatures. Box is made of triple density expanded polystyrene and molded under extreme pressure creating a durable container that guarantees temperature stability. Easily interchange cool chambers and cool bricks for different sized tubes.
Each complete CoolSafe System includes 1 CoolSafe box with lid, 1 Cool Chamber and 2 Cool Bricks (1 degree C & -10 degree C).
Capacity: 32