Learn More
Bio Rad Laboratories COOMASIE BR.BLUE R250 10G
Catalog No. NC9344796
Encompass
Coomassie Brilliant Blue R-250 Reagent, 10g, The fastest and easiest way to Coomassie-stain Criterion or other protein polyacrylamide gels
This item is not returnable. View return policy
Catalog No. NC9344796 Supplier Bio Rad Laboratories Supplier No. 1610400
Coomassie Brilliant Blue R-250 Reagent, 10g, The fastest and easiest way to Coomassie-stain Criterion or other protein polyacrylamide gels