Learn More
Biotang Inc COOMASSIE BLUE G-250, 500 ML,ST-140, 2% Coomassie Brilliant Blue G-250 in 30% ethanol. Blue dye with slight green tint. Shipped at room temperature. Store at room temperature.
Catalog No. 50152420 Shop All BIOTANG Inc Products
Encompass_Preferred
COOMASSIE BLUE G-250, 500 ML,ST-140, 2% Coomassie Brilliant Blue G-250 in 30% ethanol. Blue dye with slight green tint. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-420 Supplier Biotang Inc Supplier No. BTBB966
COOMASSIE BLUE G-250, 500 ML,ST-140, 2% Coomassie Brilliant Blue G-250 in 30% ethanol. Blue dye with slight green tint. Shipped at room temperature. Store at room temperature.