Learn More
Alkali Scientific Coomassie Blue R 250, 100 G
SDP
Catalog No. 5000660344
Encompass_Preferred
Coomassie Blue R250 (100g) is an essential protein stain used in laboratories for protein electrophoresis, Western blotting, and gel analysis. This large 100g container is ideal for high-throughput laboratories or institutions that require significant amounts of stain for routine protein analysis. Coomassie Blue R250 effectively stains proteins by binding to the molecules, creating clear, visible bands that can be analyzed under gel imaging systems. This dye is widely utilized in molecular biology and biochemistry for detecting proteins in various types of electrophoresis, including SDS-PAGE. The 100g size ensures that labs have a sufficient supply for multiple experiments and protein assays.
Catalog No. 50-006-60344 Supplier Alkali Scientific Supplier No. COO003CP
Coomassie Blue R250 (100g) is an essential protein stain used in laboratories for protein electrophoresis, Western blotting, and gel analysis. This large 100g container is ideal for high-throughput laboratories or institutions that require significant amounts of stain for routine protein analysis. Coomassie Blue R250 effectively stains proteins by binding to the molecules, creating clear, visible bands that can be analyzed under gel imaging systems. This dye is widely utilized in molecular biology and biochemistry for detecting proteins in various types of electrophoresis, including SDS-PAGE. The 100g size ensures that labs have a sufficient supply for multiple experiments and protein assays.