Learn More
Alkali Scientific Coomassie Blue R 250, 25 Grams
SDP
Catalog No. 5000660687
Encompass_Preferred
Coomassie Blue R250, 25g is a well-known dye used in protein staining, particularly in SDS-PAGE gel electrophoresis for visualizing protein bands. The dye binds to proteins, creating a blue stain that can be visualized under the appropriate light source. Coomassie Blue R250 is widely used in laboratories for protein quantification and molecular biology applications. This 25g pack size is suitable for laboratories with moderate-scale requirements for protein analysis. It is commonly used in conjunction with other laboratory techniques like Western blotting and for assessing protein purity and quantity in research studies. Coomassie Blue R250 offers a simple, effective way to monitor protein separation and expression.
Catalog No. 50-006-60687 Supplier Alkali Scientific Supplier No. COO001CP
Coomassie Blue R250, 25g is a well-known dye used in protein staining, particularly in SDS-PAGE gel electrophoresis for visualizing protein bands. The dye binds to proteins, creating a blue stain that can be visualized under the appropriate light source. Coomassie Blue R250 is widely used in laboratories for protein quantification and molecular biology applications. This 25g pack size is suitable for laboratories with moderate-scale requirements for protein analysis. It is commonly used in conjunction with other laboratory techniques like Western blotting and for assessing protein purity and quantity in research studies. Coomassie Blue R250 offers a simple, effective way to monitor protein separation and expression.