Learn More
Bioworld Coomassie Blue R-250 (2X), 1 L
Catalog No. 50255824
Encompass_Preferred
Stain for location and detection of proteins. Protocol: After SDS-PAGE, transfer the gels in fixing solution (30% methanol and 7% acetic acid) for 30 min. under constant agitation. Replace the fixing solution, submerge the gel in stain and shake the gel gently gor 2-4 hrs. Pour off the stain, rinse the gel briefly with fixing solution and cover the gel with fresh fixing solution for 30 min. Repeat and continue destaining until distinct blue bands against a clear background are obtained. Store the gel in 7% acetic acid solution. Photograph or dry the gel according to your standard protocol.Synonym: Brilliant blue R; Coomassie brilliant blue R-250; Acid Blue 83; CI 42660Appearance: Liquid
Catalog No. 50-255-824 Supplier Bioworld Supplier No. 403400042
Stain for location and detection of proteins. Protocol: After SDS-PAGE, transfer the gels in fixing solution (30% methanol and 7% acetic acid) for 30 min. under constant agitation. Replace the fixing solution, submerge the gel in stain and shake the gel gently gor 2-4 hrs. Pour off the stain, rinse the gel briefly with fixing solution and cover the gel with fresh fixing solution for 30 min. Repeat and continue destaining until distinct blue bands against a clear background are obtained. Store the gel in 7% acetic acid solution. Photograph or dry the gel according to your standard protocol.Synonym: Brilliant blue R; Coomassie brilliant blue R-250; Acid Blue 83; CI 42660Appearance: Liquid