Learn More
Alkali Scientific Coomassie Blue R 250, 500 G
SDP
Supplier:  Alkali Scientific COO004CP
Encompass_Preferred
Coomassie Blue R250 (500g) is a highly concentrated protein stain used in electrophoresis, Western blotting, and protein gel analysis. This 500g bulk quantity is ideal for large-scale protein analysis and high-throughput laboratories that require frequent staining for multiple experiments. Coomassie Blue R250 is a widely used dye in protein chemistry because of its sensitivity and ability to generate vivid, clear protein bands for easy visualization. The dye binds to proteins in polyacrylamide gels and is commonly used in molecular biology and biochemistry to determine protein concentrations, identify protein markers, and confirm the results of protein electrophoresis experiments.
Catalog No. 50-006-60721