Learn More
Biotang Inc COOMASSIE BLUE R-250, 500 ML,ST-120, 2% Coomassie Brilliant Blue R-250 in 30% ethanol. Blue dye with slight red tint. Shipped at room temperature. Store at room temperature.
Catalog No. 50152419 Shop All BIOTANG Inc Products
Encompass_Preferred
COOMASSIE BLUE R-250, 500 ML,ST-120, 2% Coomassie Brilliant Blue R-250 in 30% ethanol. Blue dye with slight red tint. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-419 Supplier Biotang Inc Supplier No. BTBB965
COOMASSIE BLUE R-250, 500 ML,ST-120, 2% Coomassie Brilliant Blue R-250 in 30% ethanol. Blue dye with slight red tint. Shipped at room temperature. Store at room temperature.