Learn More
Teknova Coomassie Blue R-250 Stain. 1000mL, Non-Sterile. 8 Pack.
SDP
Catalog No. 502387764
Encompass
0.05% Coomassie Blue R-250, 50.00% Methyl Alcohol, 10.00% Acetic Acid
Catalog No. 50-238-7764 Supplier Teknova Supplier No. C10508PK
0.05% Coomassie Blue R-250, 50.00% Methyl Alcohol, 10.00% Acetic Acid