Learn More
Bioworld Coomassie Brilliant Blue G-250, 25 g
Catalog No. 50255826
Encompass_Preferred
Protein electrophoresis dye used in SDS-PAGE.Also used for protein concentration determination by Bradford method.A commonly used analytical biochemistry dye and P2X7 antagonistUltra Pure Grade.Average purity greater than 85%.EM (595nm, water) >36,300Protein Staining: PassesAppearance: SolidMelting Point: >100 °C
Catalog No. 50-255-826 Supplier Bioworld Supplier No. 403400082
Protein electrophoresis dye used in SDS-PAGE.Also used for protein concentration determination by Bradford method.A commonly used analytical biochemistry dye and P2X7 antagonistUltra Pure Grade.Average purity greater than 85%.EM (595nm, water) >36,300Protein Staining: PassesAppearance: SolidMelting Point: >100 °C