Learn More
Gold Biotechnology Inc Coomassie Brilliant Blue G 250
SDP
Catalog No. 5000562052
Encompass_Preferred
Coomassie Brilliant Blue G-250 (CBBG) is a member of the Coomassie family and is used as a common analytical dye in SDS-PAGE and BN-PAGE CBBG can also be used with the Bradford Method to determine protein concentration
Catalog No. 50-005-62052 Supplier Gold Biotechnology Inc Supplier No. C46025
Coomassie Brilliant Blue G-250 (CBBG) is a member of the Coomassie family and is used as a common analytical dye in SDS-PAGE and BN-PAGE CBBG can also be used with the Bradford Method to determine protein concentration