Learn More
Bioworld Coomassie Brilliant Blue R-250, 25 g
Catalog No. 50255829
Encompass_Preferred
Protein electrophoresis dye used in SDS-PAGE.More sensitive than G-250.Ultra Pure GradeEM (550nm, water) >33,000Protein Staining: PassesAppearance: Solid
Catalog No. 50-255-829 Supplier Bioworld Supplier No. 403400122
Protein electrophoresis dye used in SDS-PAGE.More sensitive than G-250.Ultra Pure GradeEM (550nm, water) >33,000Protein Staining: PassesAppearance: Solid