Learn More
Abcam Coomassie Brilliant Blue R-250 Protein Stain, 25G
Catalog No. 502704789
Encompass_Preferred
MW 825.99 g/mol. Triphenylmethane dye used in SDS-PAGE for the analysis of proteins. The dye molecule binds to proteins to form a protein-dye complex. This complex stabilizes the negatively charged anionic form of the dye producing the blue color, even under acidic conditions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4789 Supplier Abcam Supplier No. AB14626025G
MW 825.99 g/mol. Triphenylmethane dye used in SDS-PAGE for the analysis of proteins. The dye molecule binds to proteins to form a protein-dye complex. This complex stabilizes the negatively charged anionic form of the dye producing the blue color, even under acidic conditions.
The product is subject to the following: Abcam Restricted Use Statement