Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coomassie Brilliant Blue R-250, Electrophoresis Grade, MP Biomedicals™

Catalog No. 04821624 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-821-624 10 g
04-821-616 25 g
04-821-636 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 04-821-624 Supplier MP Biomedicals, Inc Supplier No. 04821624
Add to Cart
Add to Cart

Coomassie Brilliant Blue R-250, Electrophoresis Grade stain is for use in protein gel staining.

  • Brilliant Blue R can be used for staining proteins on polyacrylamide electrophoresis gels.
  • Improvement of sensitivity and background have been accomplished using Brilliant Blue dyes in a colloidal form.
  • Other applications for brilliant Blue R include use in detection of lipids in thin layer chromatography, determination of the CMC of detergents, and detection of proteins in solution, as well as on cellulose acetate plates.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6104-59-2
Form Powder
Molecular Weight (g/mol) 825.971 g/mol
Purity or Quality Grade Electrophoresis
Quantity 10 g
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.