Learn More
Newcomer Supply Copper, Rhodanine Stain Kit, 250 ml kit
SDP
Catalog No. NC0831263
Encompass
Newcomer Supply Copper, Rhodanine Stain Kit procedure, with included microwave modification, is used for detection of copper and copper-associated protein (CAP) in tissue sections. Abnormal copper accumulations are predominantly found in liver tissue, most notably in Wilson’s disease.
Catalog No. NC0831263 Supplier Newcomer Supply Supplier No. 9113A
Newcomer Supply Copper, Rhodanine Stain Kit procedure, with included microwave modification, is used for detection of copper and copper-associated protein (CAP) in tissue sections. Abnormal copper accumulations are predominantly found in liver tissue, most notably in Wilson’s disease.