Learn More
Electron Microscopy Sciences Embra Coordinator 100 mesh Grid, Nickel
SDP
Catalog No. 5025117
Encompass_Preferred
Standard 3.05 mm diameter grids with a handle.
Grid: Nickel
Catalog No. 50-251-17 Supplier Electron Microscopy Sciences Supplier No. EC100NI
Standard 3.05 mm diameter grids with a handle.
Grid: Nickel