Learn More
Electron Microscopy Sciences Embra Coordinator 200 mesh Grid, Nickel
SDP
Catalog No. 5025119
Encompass_Preferred
Standard 3.05 mm diameter grids with a handle.
Grid: Nickel
Catalog No. 50-251-19 Supplier Electron Microscopy Sciences Supplier No. EC200NI
Standard 3.05 mm diameter grids with a handle.
Grid: Nickel