Learn More
CoorsTek™ AD-998 High-Alumina Disc
Catalog No. 0822617G
Change view
Click to view available options
Diameter (Metric):
102 mm
Catalog No. Diameter (Metric)
08-226-17G 102 mm
1 options
Catalog No. 08-226-17G Supplier CoorsTek™ Supplier No. 65637
Specifications
Material Alumina
Type Disc
For Use With (Application) For refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium
Diameter (English) 4 in.
Diameter (Metric) 102 mm