Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM Buchner funnel cap1EA
Catalog No. 502900918
Encompass
Porcelain with fixed perforated plate. Glazed inside and out except for rim.
Catalog No. 50-290-0918 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2473591EA
Porcelain with fixed perforated plate. Glazed inside and out except for rim.