Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM Buchner funnel cap1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2473161EA
Encompass
Porcelain with fixed perforated plate. Glazed inside and out except for rim.
Catalog No. 50-290-1745