Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM Gooch crucible por1EA
Catalog No. 502901746
Encompass
Glazed inside and out except for outside bottom surface and rim. Used for total suspended solids determinations.
Catalog No. 50-290-1746 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2470731EA
Glazed inside and out except for outside bottom surface and rim. Used for total suspended solids determinations.