Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM high-alumina cruci1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z2476261EA
Made of 99.8% pure recrystallized aluminum oxide for high resistance to chemical attack. for use in reducing and oxidizing atmospheres and refractory metals (such as molybdenum platinum rhodium tungsten tantalum and iridium). O.D. dimension tolerance is /-5%. Max. use temp. (no load) 1 750 C.