Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM porcelain crucible
Catalog No. 502901747
Encompass
Glazed inside and out except for outside bottom surface and rim.Max. use temp. 1150C limit of use evacuated 1300CCrucibles are not porous so there should not be any change in weight after firingOrder covers separately.
Catalog No. 50-290-1747 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2471031PAK
Glazed inside and out except for outside bottom surface and rim.Max. use temp. 1150C limit of use evacuated 1300CCrucibles are not porous so there should not be any change in weight after firingOrder covers separately.