Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM porcelain crucible
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2471381PAK
Encompass
Glazed inside and out except for outside bottom surface and rim.Max. use temp. 1150C limit of use evacuated 1300CCrucibles are not porous so there should not be any change in weight after firingOrder covers separately.
Catalog No. 50-290-1693