Learn More
Sigma Aldrich Fine Chemicals Biosciences CoorsTM porcelain mortar O1EA
Catalog No. 502900408
Encompass
Mortar with lip glazed outside except for bottom. Order pestle separately.
Catalog No. 50-290-0408 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2474991EA
Mortar with lip glazed outside except for bottom. Order pestle separately.