Learn More
Enzo Life Sciences COP9 signalosome Csn6 subunit (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82950025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 283-297 of human COP9 signalosome (Csn6 subunit). Source: Purified from rabbit serum. Species reactivity: Human. Specificity: Recognizes the Csn6 subunit of the COP9 signalosome. Applications: WB. Purity: Partially purified immunoglobulin. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1617